missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ IC183B TS Coverall, Sterile, Double Bagged

Beskrivning
- Gamma-sterilized
- Bound internal seams
- Elasticated waist at back for good fit
- Tyvek™ covered elasticated thumb loops
- Tunneled elastication at wrists and ankles
- Front zipper closure
- Storm flap
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms, and non-hazardous water-based light liquid splash
- Breathable and low linting
- Sterilized by gamma-irradiation
- Folded to aid aseptic donning and packaged in an ISO class 5 cleanroom
- Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag
- Packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
- Typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments
Specifikationer
Specifikationer
| Product Type | Cleanroom Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?