Learn More
DuPont™ Tyvek™ IsoClean™ IC193B TS Coverall, Sterile, Double Bagged
BIOPHARMA
Product Code. 17839413
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17839413 X-Large Pack of 20
17809413 Small Pack of 20
17899403 Medium Pack of 20
17889403 Large Pack of 20
17879413 2X-Large Pack of 20
17869413 3X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17839413 Supplier DuPont™ Supplier No. D15560746
  • ONLINE EXCLUSIVE PRICE
    475.74 €
    300.70 € / Pack of 20
    Save 175.04 €
    36% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hooded gamma-sterilized coveralls are aseptically folded and feature bound internal seams, integrated overboots with a slip-retardant sole, tunnelled elastication at wrists and ankles, elasticated waist at back, Tyvek™ covered elasticated thumb loops, and a storm flap.
  • Gamma-sterilized
  • Bound internal seams
  • Elasticated waist at back for good fit
  • Tyvek™ covered elasticated thumb loops
  • Tunneled elastication at wrists and ankles
  • Front zipper closure
  • Storm flap
  • Delivers an ideal balance of protection, durability and comfort
  • Made of high density polyethylene using a patented flash spinning process
  • Provides an inherent barrier to particles, microorganisms, and non-hazardous water-based light liquid splash
  • Breathable and low linting
  • Sterilized by gamma-irradiation
  • Folded to aid aseptic donning and packaged in an ISO class 5 cleanroom
  • Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag
  • Packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
  • Typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Finger Loops
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Particle
Latex Free Yes
Model D15560