missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ IC501B WS Sleeve, Sterile, Double Bagged
BIOPHARMA

Product Code. 17839323
Change view
Click to view available options
Length (Metric):
45 cm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
17839323 45 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17839323 Supplier DuPont™ Supplier No. D15560753
  • ONLINE EXCLUSIVE PRICE
    216.53 €
    139.42 € / Pack of 100
    Save 77.11 €
    35% Off
View all promotions
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gamma-sterilized sleeve is asepcially folded and features bound internal seams, and tunnelled elastication at wrist and bicep.

  • 45 cm long sleeve available in white and in one size
  • Gamma-sterilized
  • Bound seams
  • Tunnelled elastication at wrist and bicep.

Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.

Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.

Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.

TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Construction Gamma-sterilized, bound internal seams, tunnelled elastication at wrist and bicep, aseptically folded
Product Type Controlled Environment Sleeve
Material Tyvek
Color White
Sterility Sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Bound
Latex Free Yes
Model D15560753
Length (Metric) 45 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.