missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 500 Laboratory Coats Press Studs

Product Code. 11752594 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 10
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11752594 2X-Large Pack of 50
11742594 Large Pack of 50
15541362 Medium Pack of 10
15531362 X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11752594 Supplier DuPont™ Supplier No. D13396040
  • ONLINE EXCLUSIVE PRICE
    403.91 €
    305.09 € / Pack of 50
    Save 98.82 €
    24% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers enhanced protection for body parts that are more exposed to hazardous substances, fine particles, and fibres. Other features include ultra low-linting and antistatically treated.

  • Silicon non-added
  • Stitched internal seams
  • 3 pockets and 5 press stud closures
  • Composed of flash spun high density polyethylene
  • Provides an idea balance of protection, durability and comfort
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Size 2X-Large
Gender Unisex
Material HDPE
Color White
Closure Type Press Studs
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
No. of Pockets 3
Seam Type Stitched, Internal
Sleeve Style Straight
Certifications/Compliance PPE Category, Antistatic treatment (EN 1149-5) - on both sides
For Use With (Application) Pharmaceutical, Lead and Asbestos Abatement/Remediation, General Maintenancce/Operations, Spray Painting, Clean-up
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.