missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ Collared Laboratory Coats with Press Studs, No Pockets

Product Code. 12392259 Shop All DuPont Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
Unit Size:
Pack of 10
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12392259 Large Pack of 10
11733123 2X-Large Pack of 50
12302269 X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12392259 Supplier DuPont™ Supplier No. D13398884
  • ONLINE EXCLUSIVE PRICE
    75.84 €
    54.62 € / Pack of 10
    Save 21.22 €
    27% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coat is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.

  • Silicon non-added.
  • With 5 press stud closures.
  • Elasticated cuffs (not tunnelled).
  • Stitched internal seams.
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols.
  • Excellent barrier against fine particles and fibers (down to 1 micron in size).
  • Ultra-low-linting.
  • Antistatically treated.
  • Applications include:
    • Pharmaceutical Handling
    • Chemical Protection
    • Lead and Asbestos Abatement/Remediation
    • General Maintenance/Operations
    • Spray Painting and General Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Size Large
Gender Unisex
Material HDPE
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Style Below Hip-length
No. of Pockets None
Seam Type Stitched, Internal
Certifications/Compliance EN 1149-5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.