Learn More
DuPont™ Tyvek 500 Trousers
Product Code. 12603700 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Each
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12603700 Medium Pack of 50
12912219 X-Large -
12623690 X-Large Pack of 50
11743123 Large Pack of 50
12992209 Large -
12693690 2X-Large Pack of 50
12922219 2X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12603700 Supplier DuPont™ Supplier No. D13496109
  • ONLINE EXCLUSIVE PRICE
    282.11 €
    243.89 € / Pack of 50
    Save 38.22 €
    13% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ 500 Trousers offer an excellent barrier against fine particles and fibers down to 1 micron in size.
  • Flash-spun HDPE construction
  • Elasticated waist
  • Stitched seams
  • No pockets
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Flashspun HDPE
Color White
Certifications/Compliance CE Certified, EN 1149-5
Waist Style Elastic
For Use With (Application) Applications includes pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Product Type Trousers