Learn More
DuPont™ Tyvek™ Tychem™ PL50 Laboratory Gowns
Product Code. 12613690
Change view
Click to view available options
Size: Large to XXL
Product Code.
12613690 Size: Large to XXL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12613690 Supplier DuPont™ Supplier No. D13984782
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ Tychem™ PL50 Laboratory Gowns are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek substrate.
  • Elasticated wrists
  • Antistatic treatment
  • With wrap-over rear closure, hook and loop neck closure and waist ties
  • Excellent chemical permeation protection to an extensive range of chemicals
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications
Size Large/X-Large
Gender Unisex
No. of Pockets None
Style Shin Length
Product Type General Purpose Gown
Material Tychem
Color Gray
Closure Type Wrap-around Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Seam Type Stitched and overlapped
Autoclavable Not Autoclavable