missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U-0521, 98%

Product Code. 15408719
Change view
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408719 20 mg 20mg
15418719 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408719 Supplier Thermo Scientific Chemicals Supplier No. J67355.F+
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-89-7
Molecular Formula C10H12O3
Synonym 3',4'-Dihydroxy-2-methylpropiophenone;

Specifications

Quantity 20 mg
Description White
Formula Weight 180.20
Melting Point 79°C to 81°C
Product Type U-0521, 98%
Form Solid
InChI Key VDQLKIBLTMPAHI-UHFFFAOYSA-N
SMILES CC(C)C(=O)C1=CC=C(O)C(O)=C1
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
Molecular Weight (g/mol) 180.20
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.