missing translation for 'onlineSavingsMsg'
Learn More

U-0521, TRC

Product Code. 30133509
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133509 50 mg 50mg
30132577 100 mg 100mg
30008676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133509 Supplier Toronto Research Chemicals Supplier No. TRCU85032550MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-89-7
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.201
Synonym 3',4'-Dihydroxy-2-methylpropiophenone, 1-(3,4-Dihydroxyphenyl)-2-methyl-1-propanone, 1-(3,4-Dihydroxyphenyl)-2-methylpropanone, 3',4'-Dihydroxy-α-methylpropiophenone, NSC 27389, U 0521
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-methylpropan-1-one
SMILES CC(C)C(=O)c1ccc(O)c(O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12O3/c1-6(2)10(13)7-3-4-8(11)9(12)5-7/h3-6,11-12H,1-2H3
Formula Weight 180.079
Chemical Name or Material U-0521
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.