missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ U 73122

Product Code. 30001221
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001221 10 mg 10mg
18014352 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001221 Supplier R&D Systems™ Supplier No. 1268/10
  • ONLINE EXCLUSIVE PRICE
    247.59 €
    222.60 € / 10mg
    Save 24.99 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Phospholipase C inhibitor

Phospholipase C inhibitor. Inhibits agonist-induced platelet aggregation with IC50 values of 1-5 μM. Potently inhibits human polymorphonuclear neutrophil adhesion on biological surfaces (IC50 < 50 nM) and exhibits antinociceptive activity in vivo. Also activates TRPM4 and inhibits TRPM3 channels.

Negative Control also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112648-68-7
Molecular Formula C29H40N2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 464.65
Purity >98%
Molecular Weight (g/mol) 464.65
Chemical Name or Material U 73122
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.