Learn More
Thermo Scientific Chemicals U-73122, 95%
Description
It is used as a phospholipase C, phospholipase A2, and 5-LO inhibitor. It is also determined that in SK-N-SH neuroblastoma cells, U-73122 inhibits agonist-induced down-regulation of muscarinic receptors. In addition, it is a useful tool to investigate receptor-mediated PI turnover in signal transduction. U-73122 is a potent inhibitor of human neutrophil adhesion to biological surfaces (IC50 = 50 nM) as well as adhesion-dependent granule exocytosis and oxidative burst. U-73343 (sc-201422) is useful as a negative control for investigations of U-73122 phospholipase C antagonism and its cellular consequences.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 mg |
| Description | Off-white |
| For Use With (Application) | Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors |
| Formula Weight | 464.65g/mol |
| Solubility | Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml; May require heating or overnight mixing; Insoluble in water |
| Product Type | Signal Transduction Reagent |
| Form | Crystalline solid |
| Assay | ≥99% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.