missing translation for 'onlineSavingsMsg'
Learn More

U-73122, MedChemExpress

Product Code. 16481837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481837 50 mg 50mg
16468367 5 mg 5mg
16415678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481837 Supplier Medchem Express Supplier No. HY13419
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50of 1-2.1 μM for PLC.

U-73122 is a phospholipase C (PLC) and 5-LO (5-lipoxygenase) inhibitor with an IC50of 1-2.1 μM for PLC.

Specifications

Chemical Name or Material U-73122
CAS 112648-68-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H40N2O3
Quantity 50 mg
Solubility Information DMSO : 12.5 mg/mL (26.90 mM; Need ultrasonic)
SMILES O=C(C=C1)N(CCCCCCN[C@H]2CC[C@@]3([H])[C@]4([H])CCC5=C(C=CC(OC)=C5)[C@@]4([H])CC[C@]23C)C1=O
Molecular Weight (g/mol) 464.64
Formula Weight 464.64
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.