missing translation for 'onlineSavingsMsg'
Learn More

uvex U-Chem 3100 Chemical Protection Gloves

Product Code. 17240521 Shop All Uvex Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17240521 8 10 pairs
17250521 9 10 pairs
17260521 10 10 pairs
17717639 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17240521 Supplier uvex Supplier No. 6096808
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are constructed of nitrile butadiene rubber (NBR) with a cotton interlock liner, an additional coating for enhances grip capabilities when handling wet or oily objects.

  • Nitrile butadiene rubber (NBR) chemical protection gloves
  • An additional coating enhances grip capabilities when handling wet/oily objects
  • Ideal for a wide variety of applications that require mechanical protection
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Black
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 4121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMO
For Use With (Application) Automotive, Chemical Handling, Metal Work, Mechanics, and Sand Blasting
Gender Unisex
Glove Cuff Style Gauntlet
Series U-Chem 3100
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.