Learn More
uvex u-fit Safety Gloves
Product Code. 31903211 Shop All Uvex Products
Change view
Click to view available options
Size:
6, X-Small
7, Small
8, Medium
9, Large
10, X-Large
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31903211 10, X-Large Each
31903203 6, X-Small Each
31903167 7, Small Each
31903181 8, Medium Each
31903178 9, Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31903211 Supplier uvex Supplier No. 6072710
  • ONLINE EXCLUSIVE PRICE
    9.82 €
    9.01 € / Pack of 1
    Save 0.81 €
    8% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The gloves offer reliable protection and comfortable, precise working in various applications such as chemistry, medicine, the food industry and production.
  • The u-fit series of high-quality disposable safety gloves are made of NBR and ensure a high level of safety and functionality.
TRUSTED_SUSTAINABILITY
Specifications
Applications Healthcare, Laboratory, Industrial
Latex Free Yes
Material Nitrile
Product Type Disposable Gloves
Size 10, X-Large