missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The u-fit series of high-quality disposable safety gloves are made of NBR and ensure a high level of safety and functionality.
Specifications
Specifications
| Applications | Healthcare, Laboratory, Industrial |
| Color | Blue |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Disposable Gloves |
| Size | 6, X-Small |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?