missing translation for 'onlineSavingsMsg'
Learn More

uvex U-Fit Strong N2000 Chemical Protection Gloves

Product Code. 17200521 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17200521 Medium Each
17290511 Small Each
17210521 Large Each
17220521 X-Large Each
17230521 2X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17200521 Supplier uvex Supplier No. 6096208
  • ONLINE EXCLUSIVE PRICE
    29.00 €
    17.89 € / Pack of 1
    Save 11.11 €
    38% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reinforced, disposable nitrile rubber safety gloves provide protection against chemicals and splashes, with a high degree of mechanical resistance.

  • Reinforced, disposable nitrile rubber safety gloves
  • Textured fingertips for enhanced grip capabilities
  • Resistant to grease, mineral oils, and a wide variety of chemicals
  • Silicone-free according to imprint test
  • Ideal for cleaning, laboratory work, and the food industry
TRUSTED_SUSTAINABILITY

Specifica

Applications Laboratory
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile Rubber
Product Type Chemical-resistant Gloves
Size Medium
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPST
For Use With (Application) Chemical Handling, Cleaning, Food Industry, Laboratory Work, and Painting
Gender Unisex
Series U-Fit Strong N2000
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.