missing translation for 'onlineSavingsMsg'
Learn More

uvex U-Fit Strong N2000 Chemical Protection Gloves

Product Code. 17290511 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17290511 Small Each
17200521 Medium Each
17210521 Large Each
17220521 X-Large Each
17230521 2X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17290511 Supplier uvex Supplier No. 6096207
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Reinforced, disposable nitrile rubber safety gloves provide protection against chemicals and splashes, with a high degree of mechanical resistance.

  • Reinforced, disposable nitrile rubber safety gloves
  • Textured fingertips for enhanced grip capabilities
  • Resistant to grease, mineral oils, and a wide variety of chemicals
  • Silicone-free according to imprint test
  • Ideal for cleaning, laboratory work, and the food industry
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile Rubber
Product Type Chemical-resistant Gloves
Size Small
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPST
For Use With (Application) Chemical Handling, Cleaning, Food Industry, Laboratory Work, and Painting
Gender Unisex
Series U-Fit Strong N2000
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.