missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ U0126

Product Code. 30001399
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001399 25 mg 25mg
30001208 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001399 Supplier R&D Systems™ Supplier No. 1144/25
  • ONLINE EXCLUSIVE PRICE
    1324.92 €
    1191.75 € / 25mg
    Save 133.17 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective inhibitor of MEK1 and 2

Potent and selective non-competitive inhibitor of MAP kinase kinase. Inhibits MEK-1 and MEK-2 (IC50 values of 0.07 and 0.06 μM respectively) with little or no effect on the activities of PKC, Abl, Raf, MEKK, ERK, JNK, MKK-3, MKK-4/SEK, MKK-6, Cdk2 or Cdk4. Brain penetrant. Helps maintain human pluripotent stem cells in undifferentiated state.

Negative Control also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109511-58-2
Molecular Formula C18H16N6S2

Specifications

Quantity 25 mg
Product Line Tocris
Formula Weight 380.48
Purity >98%
Molecular Weight (g/mol) 380.49
Chemical Name or Material U0126
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.