missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U0126, 99+%

Product Code. 15423859
Change view
Click to view available options
Quantity:
25 mg
100 mg
300 mg
Unit Size:
100mg
25mg
300mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423859 100 mg 100mg
15413859 25 mg 25mg
15403859 300 mg 300mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423859 Supplier Thermo Scientific Chemicals Supplier No. J61246.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2 | CAS: 109511-58-2 | C18H16N6S2, C18H16N6S2 | 380.49 g/mol

U0126, is used as a selective MAP Kinase Kinase (MKK, MEK) inhibitor. U-0126 has been observed to inhibit promoters containing an AP-1 response element, while in contrast having no effect on promoters that lack an AP-1 response group. U-0126 is also useful in neuronal studies in mice since it displays inhibitory effects against oxidative stress. U-0126 is an activator of PGC-1, mtTFA and nuclear respiratory factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109511-58-2
Molecular Formula C18H16N6S2
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene

Specifications

Quantity 100 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2
Formula Weight 380.49g/mol
Solubility Soluble in DMSO; Poorly soluble in ethanol and water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
What is U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) soluble in?

U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) is soluble in DMSO. It is poorly soluble in ethanol and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.