missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90

Product Code. 15924426 Shop All asecos Products
Change view
Click to view available options
Door Style:
Sliding
Model:
UB90.060.059.050.S
UB90.060.059.S
UB90.060.089.050.S
UB90.060.089.S
UB90.060.110.050.2S
UB90.060.110.050.S
UB90.060.110.2S
UB90.060.110.S
UB90.060.140.2S
UB90.080.059.060.S
UB90.080.110.060.2S
Weight (Metric):
111 kg
123.30 kg
148 kg
150.50 kg
163 kg
180 kg
183.50 kg
193 kg
200.10 kg
244 kg
244.50 kg
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Model Weight (Metric) Unit Size
15924426 Sliding UB90.060.110.S 193 kg Each
15974526 Sliding UB90.060.059.050.S 111 kg Each
15924506 Sliding UB90.060.089.050.S 148 kg Each
15904436 Sliding UB90.060.110.050.S 180 kg Each
15944456 Sliding UB90.060.110.050.2S 183.50 kg Each
15958062 Sliding UB90.060.059.S 123.30 kg Each
15904496 Sliding UB90.060.089.S 163 kg Each
15994436 Sliding UB90.060.110.2S 200.10 kg Each
15974396 Sliding UB90.060.140.2S 244 kg Each
15927476 Sliding UB90.080.059.060.S 150.50 kg Each
15907476 Sliding UB90.080.110.060.2S 244.50 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15924426 Supplier asecos™ Supplier No. 3042000135327

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly: drawers remain open in any position, interior of drawers completely visible – access to all containers
  • Safe: drawers fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: drawers lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Warranty

Five years

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 1 x drawer (sheet steel powder-coated), 1 x perforated insert (stainless steel)
Depth (Metric) Exterior 574 mm
Depth (Metric) Interior 452 mm
Dimensions (D x W x H) Interior 452 x 979 x 502 mm
Dimensions (D x W x H) 574 x 1102 x 600 mm
Door Style Sliding
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 600 mm
Height (Metric) Interior 502 mm
Model UB90.060.110.S
No. of Doors 1
Surface Coating Powder-coated plain
Weight (Metric) 193 kg
Width (Metric) Exterior 1102 mm
Width (Metric) Interior 979 mm
Color Light Gray
Material Steel
Product Type Safety Storage Cabinet (Under Bench)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.