missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet

Product Code. 15536644 Shop All asecos Products
Click to view available options
Weight (English):
364.42 lb.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15536644

Brand: asecos™ 3044400133775

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Safe and approved storage of flammable hazardous substances in working areas with easy moving of the cabinet

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Specifications

Certifications/Compliance DIN EN 16121/16122 conformity, GS Certified, CE Compliant
Configuration 1 x Perforated Insert, 1 x Bottom Collection Sump
Depth (English) Interior 15.04 in.
Depth (Metric) Exterior 502 mm
Depth (Metric) Interior 382 mm
Dimensions (D x W x H) Interior 382 x 979 x 502 mm
Dimensions (D x W x H) 502 x 1102 x 600 mm
Door Style Wing
For Use With (Application) Storage of Flammable Hazardous Substances
Height (English) Interior 19.76 in.
Height (Metric) Exterior 600 mm
Height (Metric) Interior 502 mm
Includes Under Bench Cabinet with Interior Fittings
Load Bearing Capacity Insert 75 kg, Sump 21 L, Maximum load: 300 kg
Model UB90.060.110.050.2T
No. of Doors 2
No. of Shelves 0
Surface Coating Powder-Coated Plain
Weight (English) 364.42 lb.
Weight (Metric) 165.30 kg
Width (English) Interior 38.54 in.
Width (Metric) Exterior 1102 mm
Width (Metric) Interior 979 mm
Color Light Gray
Depth (English) Exterior 19.76 in.
Height (English) Exterior 23.62 in.
Height (Metric) Opening 502 mm
Material Sheet Steel
Product Type Flammables Storage Cabinet
Width (English) Exterior 43.39 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.