Learn More
Poulten Selfe™ Ubbelohde Suspended-Level Viscometers
Product Code. 13141063
Change view
Click to view available options
Size:
1B
1C
2
2C
3B
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
13141063 1B Each
13151063 1C Each
13171063 2 Each
13181063 2C Each
13121073 3B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13141063 Supplier Poulten Selfe™ Supplier No. 1643/04
Suitable for transparent liquids. With UKAS certificate of calibration
ASTM D445-IP71, ASTM D446.Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.
Specifications
Product Type Ubbelohde Suspended-Level Viscometer
Viscosity Range 2 to 10 cSt
ASTM Numbers D445-IP71, D446
Certifications/Compliance D446
Size 1B
Volume (Metric) 18 mL