Learn More
Paragon Scientific™ Ubbelohde Viscometer for Tranparent Liquids
Product Code. 16040470
Change view
Click to view available options
Viscosity Range:
0.3 to 1 cSt
0.6 to 3 cSt
1 to 5 cSt
10 to 50 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1000 to 5000 cSt
2 to 10 cSt
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
16040470 0.3 to 1 cSt Each
16060470 0.6 to 3 cSt Each
16080470 1 to 5 cSt Each
16000480 2 to 10 cSt Each
16020480 6 to 30 cSt Each
16040480 10 to 50 cSt Each
16060480 20 to 100 cSt Each
16080480 60 to 300 cSt Each
16000490 100 to 500 cSt Each
16020490 200 to 1000 cSt Each
16040490 600 to 3000 cSt Each
16060490 1000 to 5000 cSt Each
16080490 2000 to 10,000 cSt Each
16000500 6000 to 30,000 cSt Each
16020500 10,000 to 50,000 cSt Each
16040500 20,000 to 100,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 16040470 Supplier Paragon Scientific™ Supplier No. UBBEL01NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 0.3 to 1 cSt
Calibrated No
Approximate Constant 0.001
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size 0
4 week lead-time