missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ubiquitin E1 Inhibitor, PYR-41

Product Code. 15416879
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416879 100 mg 100mg
15406879 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416879 Supplier Thermo Scientific Chemicals Supplier No. J65388.MC
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 418805-02-4 | C17H13N3O7·3H2O, C17H13N3O7 | 371.31 g/mol

PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 418805-02-4
Molecular Formula C17H13N3O7·3H2O
Synonym Ubiquitin-Activating Enzyme E1 Inhibitor; PYR-41

Specifications

Quantity 100 mg
Description Dark Purple
Content And Storage -30°C to -10°C
For Use With (Application) PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
Formula Weight 425.40g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO at 5mg/ml with warming
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.