missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ UDP disodium salt

Product Code. 30001380
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001380 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001380 Supplier R&D Systems™ Supplier No. 3111/50
  • ONLINE EXCLUSIVE PRICE
    106.40 €
    95.55 € / 50mg
    Save 10.85 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous ligand; competitive antagonist at P2Y14 receptors

Endogenous P2Y receptor agonist which preferentially activates P2Y6. Shown to be a competitive antagonist at P2Y14 receptors.
TRUSTED_SUSTAINABILITY

Specifications

CAS 27821-45-0
Quantity 50 mg
Product Line Tocris
Molecular Formula C9H12N2Na2O12P2
Formula Weight 448.12
Purity >97%
Molecular Weight (g/mol) 448.12
Chemical Name or Material UDP disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.