missing translation for 'onlineSavingsMsg'
Learn More

UDP-glucuronic Acid-13C, 15N2 Triammonium Salt, TRC

Product Code. 30008986
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
5 mg
Unit Size:
0.25mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008986 5 mg 5mg
30009433 0.25 mg 0.25mg
30012520 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008986 Supplier Toronto Research Chemicals Supplier No. TRCT8862865MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C1413CH2215N2O18P2 • 3(NH3)
Molecular Weight (g/mol) 634.35
IUPAC Name (2S,3S,4S,5R,6R)-6-(((((((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl-2-13C-1,3-15N2)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)(hydroxy)phosphoryl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid, triammonia salt
SMILES O[C@@H]1[C@@H]([C@H](O[C@H]1[15N]([13C]([15NH]2)=O)C=CC2=O)COP(O)(OP(O[C@@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(O)=O)O)O)O)(O)=O)=O)O.N.N.N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H22N2O18P2.3H3N/c18-5-1-2-17(15(26)16-5)12-9(22)6(19)4(32-12)3-31-36(27,28)35-37(29,30)34-14-10(23)7(20)8(21)11(33-14)13(24)25;;;/h1-2,4,6-12,14,19-23H,3H2,(H,24,25)(H,27,28)(H,29,30)(H,16,18,26);3*1H3/t4-,6-,7+,8+,9-,10-,11+,12-,14-;;;/m1.../s1/i15+1,16+1,17+1;;;
Formula Weight 634.12
Chemical Name or Material UDP-glucuronic Acid-13C, 15N2 Triammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.