missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT1A10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 468.00 €
Specifications
| Antigen | UGT1A10 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
UGT1A10 Polyclonal antibody specifically detects UGT1A10 in Human samples. It is validated for Western BlotSpecifications
| UGT1A10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 54575 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GNT1, UDP glucuronosyltransferase 1 family, polypeptide A10, UDP glycosyltransferase 1 family, polypeptide A10, UDP-glucuronosyltransferase 1-10, UDP-glucuronosyltransferase 1A10, UDP-glucuronosyltransferase 1-J, UDPGT, UDPGT 1-10, UGT1, UGT1*10, UGT1.10, UGT1-10, UGT-1J, UGT1JEC 2.4.1.17 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human UGT1A10 (NP_061948.1). LRGHEVVVVMPEVSWQLERSLNCTVKTYSTSYTLEDQNREFMVFAHAQWKAQAQSIFSLLMSSSSGFLDLFFSHCRSLFNDRKLVEYLKESSFDAVFLDPF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title