missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ UHPLC-MS Reagents Installation Kit
GREENER_CHOICE

Product Code. 16854941 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16854941 6 x 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16854941 Supplier Thermo Scientific™ Supplier No. UHPLCMSKIT
  • ONLINE EXCLUSIVE PRICE
    620.00 €
    601.00 € / Pack of 1
    Save 19.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

  • The installation kit includes the following reagents:
    • Two (2)  bottles of W8-1, Water, UHPLC-MS grade
    • Two (2) bottles of A458-1, Methanol, UHPLC-MS grade
    • One (1) bottle of A956-1, Acetonitrile, UHPLC-MS grade
    • One (1) bottle of T111101000, ChromaCare™ LC-MS Instrument Flush Solution
  • All reagents in the kit are packaged in 1L borosilicate glass bottles
  • By reducing background noise, the T111101000 ChromaCare™ flush solution facilitates instrument installation and preventative maintenance routines
  • All reagents in the kit are available for individual sale
TRUSTED_SUSTAINABILITY

Specifications

Quantity 6 x 1 L
Green Features ACT Ecolabel (Silver)
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • UHPLC-MS Reagents Installation Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Methanol, Acetonitrile, Isopropanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.