missing translation for 'onlineSavingsMsg'
Learn More

UK 14,304 tartrate, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30002121
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002121 10 mg 10mg
18065252 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002121 Supplier Tocris Bioscience Supplier No. 2466/10
  • ONLINE EXCLUSIVE PRICE
    215.00 €
    202.65 € / 10mg
    Save 12.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

α2 agonist. Water-soluble form of UK 14,304 (Cat. No. 0425)

Water-soluble form of UK 14,304 (Cat. No. 0425). Full α2 adrenergic agonist. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70359-46-5
Molecular Formula C15H16BrN5O6
Molecular Weight (g/mol) 442.226
InChI Key QZHBYNSSDLTCRG-YKWZRPGSSA-N
Synonym brimonidine tartrate, --tartaric acid; brimonidine
PubChem CID 72376318
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine;(2R,3S)-2,3-dihydroxybutanedioic acid
SMILES C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 442.23
Percent Purity >99%
Chemical Name or Material UK 14, 304 tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.