missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra-Pure, Chem-Lab

Product Code. 12044907
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12044907 10 L 10 L
11949242 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12044907 Supplier Chem-Lab Supplier No. CL02.2101.9010
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra purified for laboratory use, AAS and ICP. Water, Ultra-Pure, Chem-Lab meets ASTM D-94 and D-129 standards. It is UV filtrated with LF < 1μS – 0.4μm. Available in a range of quantities: 2.5, 5 and 10L.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.016g/mol
Grade Ultra Pure
Vapor Pressure 17.5mm Hg at 20°C
Filtered Through LF < 1μS – o0.4μm, UV filtrated
Density 1.00g/mL
Quantity 10 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.