missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraComp eBeads™ Spectral Unmixing Beads

Product Code. 31034433
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31034433 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31034433 Supplier Invitrogen™ Supplier No. U20250
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

UltraComp eBeads Spectral Unmixing Beads enhance spectral flow cytometry by providing precise single-color spectral unmixing control. Compatible with various antibodies and laser excitations, these third-generation beads offer optimized fluorophore unmixing, ensuring accurate results and superior performance.

UltraComp Spectral eBeads are the third generation of UltraComp beads for use in spectral flow cytometry. Fluorochrome-conjugated antibody binds the spherical particles for use as a single-color spectral unmixing control. Bind with mouse, rat, hamster, rabbit, or recombinant human antibodies.

UltraComp Spectral eBeads features include:

  • Multispecies compatibility—bind with mouse, rat, hamster, rabbit, or recombinant human antibodies
  • Improved fluorophore compatibility
  • Low frequency and severity of unmixing errors

UltraComp Spectral eBeads are designed for use in spectral unmixing and compensation with all fluorochromes excited by ultraviolet (355 nm), violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), or red (633-640 nm) lasers. These UltraComp beads have been optimized for use in spectral flow cytometry, where they unmix fluorophores in a manner similar to single color controls on cells.

Each drop of beads contains two populations: a positive population that binds with any mouse, rat, hamster, rabbit, or recombinant human antibody and a negative population that will not bind with antibody. When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed. This bimodal distribution can be used to set up single-color controls for unmixing as well as compensation in multicolor and high parameter flow cytometry experiments.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 x 106 Beads/mL
Diameter (Metric) 5 μm
Content And Storage •2°C to 8°C
•Do not freeze.
Quantity 100 Tests
Excitation Wavelength Range UV to IR
Shipping Condition Ambient
Product Type Compensation Beads
Regulatory Status RUO

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.