missing translation for 'onlineSavingsMsg'
Learn More

Advantec Disposable Ultrafiltration Unit, 200K MWCO

Product Code. 15148166 Shop All Cole-Parmer Products
Change view
Click to view available options
MWCO:
200 kDa
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
15148166 200 kDa Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15148166 Supplier Advantec Supplier No. 0291054
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Concentrate, separate, and purify your biological samples without a centrifuge using the 200K MWCO Disposable Ultrafiltration Unit.

  • Three molecular weight cutoffs.
  • 40 times maximum concentration.
  • These disposable ultrafiltration units are designed for ease in concentrating, separating, or otherwise purifying small volumes (<2 mL) of liquid samples, on the basis of molecular weight cutoff using positive pressure.
  • The unit is constructed of polypropylene cap, acrylic retentive cell, and filtrate cell.
  • Filter media is polysulfone.
  • To use, simply dispense sample into the retentive cell by pipette, place cap on retentive cell, and inject 10 mL of air with a syringe.
  • Remove cap to stop filtration.
  • Sterilization: 25% ethanol, 5% formalin

Specifications

Disposable Single-use
MWCO 200 kDa
Sterility Sterile
For Use With (Application) For use Concentrate, Separate, and Purify Biological Samples Without a Centrifuge
Quantity 24 Pk.
Type Ultrafiltration Unit
Volume (Metric) 0.5 to 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.