missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ 5M NaCl

Product Code. 17807692
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17807692 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17807692 Supplier Invitrogen™ Supplier No. A57006
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

UltraPure™ 5M NaCl is a filtered, pre-mixed solution containing 292.2 g/L NaCl. This is a common stock solution used in many molecular biology and forensic applications.

  • A filtered, pre-mixed solution containing 292.2 g/L NaCl
  • A common stock solution used in many molecular biology and forensic applications
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Chloride
Concentration 5 M
For Use With (Application) Forensics, Molecular Biology
Quantity 4 L
Purity Molecular Biology Grade
Shipping Condition Room Temperature

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.