Learn More
Cole Parmer Ultrasonic Processor Stand
Product Code. 15360538 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Equipment):
Cole-Parmer 500 -700 watt Ultrasonic Processors
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15360538 Cole-Parmer 500 -700 watt Ultrasonic Processors Each
1 options
Product Code. 15360538 Supplier Cole Parmer Supplier No. 0471292
  • ONLINE EXCLUSIVE PRICE
    146.30 €
    133.87 € / Pack of 1
    Save 12.43 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Support Stand with stainless steel rod. Enhance the performance of your ultrasonic processor.
  • Support Stand base: 9 x 5-1/2 in. 
  • Stainless Steel Rod: 24 in.H x 1/2 in. dia. 
Note: Clamp is sold separately. Order converter clamp 04714-90 for use with 130-watt ultrasonic processors. Order light-duty converter clamp 04714-91 for use with 500- and 750-watt ultrasonic processors. For industrial applications with 500- and 750-watt ultrasonic processors, order heavy-duty converter clamp 04710-94. 
Specifications
For Use With (Equipment) Cole-Parmer 500 -700 watt Ultrasonic Processors
Includes Stand and rod. (Clamp is sold separately.)
Product Type Ultrasonic Processor Stand