Learn More
uvex Ultravision Goggles with Cellulose Acetate Lens
Product Code. 17787699 Shop All Uvex Products
Change view
Click to view available options
Lens Tint: Clear
Product Code.
17787699 Lens Tint: Clear
1 options
Product Code. 17787699 Supplier uvex Supplier No. 9301906
  • ONLINE EXCLUSIVE PRICE
    16.60 €
    15.63 € / Pack of 1
    Save 0.97 €
    5% Off
View all promotions
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Uvex Ultravision features a comfortable ergonomic design and offers unrestricted peripheral vision.
These flexible goggles can also be worn as overspecs with prescription spectacles and are available with cellulose acetate or polycarbonate lenses, as welding safety spectacles or with a flip-up face guard.
General features
  • Classic safety goggle suitable for a wide range of applications
  • Indirect ventilation system increases air circulation
  • A sustainable benefit: lenses can be replaced when required
  • Available replacement lens: 9300956
  • Metal-free
Safety features
  • Certified in line with EN 166 (personal protective eyewear) and EN 170 (UV protective filter)
  • Marking: W 166 3 F CE – 2C-1,2 W1 FN CE
  • Cellulose acetate lens with anti-fog performance on the inside and UV380 protection
Comfort features
  • Flexible face seal naturally moulds to any facial shape
  • Generous internal volume accommodating prescription spectacles and frame recesses to locate the side arms ensuring a comfortable fit
  • Length-adjustable textile headband for a secure fit and individual adaptability
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Lens Coating Anti-fog
Color Transparent Gray
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Anti-fog Goggles
Headband Material Textile
Material Polycarbonate
Disposable Reusable
Lens Material Cellulose Acetate
Frame Material Plastic
UV Protection UV380
Frame Type Standard