missing translation for 'onlineSavingsMsg'
Learn More

Sartorius ULTROSER G

Product Code. 31908443 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908443 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908443 Supplier Sartorius Supplier No. 15950017
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ULTROSER G 20 mL serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium.

  • Ultroser™ G serum substitute can successfully replace FCS (fetal calf serum) in cell culture medium
  • Designed for the in vitro culture of anchorage-dependent cells
  • Features amp; BenefitsSemi-defined composition ensuring batch-to-batch reproducibility
  • Low protein content
  • Easy use and storage
  • Ultroser G serum substitute has a concentration five times higher than fetal calf serum (2 % of reconstituted Ultroser G serum substitute is equivalent to 10% fetal calf serum in the basal medium)
  • Ultroser G is for research use only and not for use in diagnostic procedures

Specifications

Country of Origin United States
Form Liquid
Format Bottle
Product Type Serum Replacement
Quantity 20 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.