missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Ultrospec™ 30 Cell Density Meter

Product Code. 30694946 Shop All Harvard Bioscience Products
Change view
Click to view available options
Range:
600 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
30694946 600 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30694946 Supplier Biochrom Supplier No. 80300812
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measures density of bacteria and yeast cells in suspension with compact and low maintenance device

  • Small, portable, and easy to use, the instrument can measure growth rates of E. coli, other bacteria, and yeast
  • Create and save cell specific standard curves to read against again and again
  • Can also be used with McFarland standards
  • Intuitive touch screen for easy plug and play use
  • Battery operated and portable for use where cells are actually cultured
  • Uses LED light source in combination with an advanced fiber optics
  • Hand - held
  • Accepts 10 x 10 mm cuvettes or 16 mm test tubes
  • 10 to 12 mm test tubes require adapter set
  • Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with formaldehyde or ethylene oxide to sterilize it
  • Save results onboard or export via USB flash drive
  • Compact, low-maintenance device
  • Not recommended for use with mammalian cell lines

Specifications

Product Type Cell Density Meter
Environmental Rating IP20
Display LED Touchscreen
Range 600 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.