Learn More
Arctiko™ ULUF P50 Compact ULT Bench Top and Undercounter Freezer
Compact ULT Integraline enables easy access to samples and provides user flexible opportunities at the bench top or under the counter.
Brand:  Arctiko™ 1000013
Product Code. 16677283
  • ONLINE EXCLUSIVE PRICE
    8333.85 €
    6745.00 € / Each
    Save 1588.85 €
    19% Off
Description
The Compact Integraline enables easy access to samples and provides user flexible opportunities in the workplace. This range of Compact Integraline freezers is ideal for placing under or on your table or counter, or near your work stations. With Arctiko™'s original single compressor technology, you are guaranteed superior temperature uniformity and outstanding cooling performance to give you time to focus on the essential: your research. The Compact ULT Freezer is an extremely reliable solution for laboratory research when your lab or clinical space is too small for a full-size ULT freezer.
  • Outstanding cooling performance with the unique single cooling system and natural refrigerant with ultra low GWP (Global Warming Potential)
  • Compact, space-saving design makes the undercounter ULT the perfect storage solution for laboratories with limited space
  • Single Compressor Technology with a simple structure ensures easy maintenance, lower heat dissipation, low energy consumption and noise reduction for a pleasant work environment
  • Direct cooling for optimized temperature stability and uniformity, even with multiple door openings
  • Heated door frame prevents ice from building up around the gasket securing a stable and uniform temperature inside the freezer
  • Low energy consumption made possible with ECO mode and ultra-efficient Vacuum Insulated Panels
  • Built-in CO2 backup controller system for extra security for your valuable samples
  • Self-diagnostic function to guarantee that your alarm system is always working correctly