Learn More
Tocris Bioscience™ UNC 1999
GREENER_CHOICE
Potent and selective EZH2/EZH1 inhibitor
Brand:  Tocris Bioscience™ 4904/50
Encompass
Product Code. 15706326
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.
Negative Control also available.