missing translation for 'onlineSavingsMsg'
Learn More

UNC2250, MedChemExpress

Product Code. p-200101394
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486838

Brand: Medchem Express HY15797

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC2250 is a potent and selective Mer inhibitor with an IC50 of 1.7 nM, about 160- and 60-fold selectivity over the closely related kinases Axl/Tyro3.

UNC2250 is a potent and selective Mer inhibitor with an IC50 of 1.7 nM, about 160- and 60-fold selectivity over the closely related kinases Axl/Tyro3.

Specifications

Chemical Name or Material UNC2250
CAS 1493694-70-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H36N6O2
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (45.39 mM; ultrasonic and warming and heat to 60°C) ∣0.1 M HCL : 12.5 mg/mL (28.37 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES CCCCNC1=NC(N[C@@H]2CC[C@@H](O)CC2)=C(C3=CC=C(CN4CCOCC4)C=N3)C=N1
Molecular Weight (g/mol) 440.58
Formula Weight 440.58
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.