missing translation for 'onlineSavingsMsg'
Learn More

gamma-Undecalactone, 98%

Product Code. 10676271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676271 25 mL Glass bottle 25mL
10792675 100 mL Glass bottle 100mL
10249873 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10676271 Supplier Thermo Scientific Acros Supplier No. 259490250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-67-6 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-67-6
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00005405
InChI Key PHXATPHONSXBIL-UHFFFAOYSA-N
Synonym gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone
PubChem CID 7714
IUPAC Name 5-heptyloxolan-2-one
SMILES CCCCCCCC1CCC(=O)O1

Specifications

Color Colorless
Density 0.9425g/mL
Boiling Point 286.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 25 mL
Specific Gravity 0.9425
Formula Weight 184.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material γ-Undecalactone
Show More Show Less
Product Identifier
  • y-Undecalactone
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.