missing translation for 'onlineSavingsMsg'
Learn More

Undecane-1,11-dicarboxylic acid, 95%, Thermo Scientific™

Product Code. 15497458
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15497458

Brand: Thermo Scientific Alfa Aesar H60962.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.331
MDL Number MFCD00002740
InChI Key DXNCZXXFRKPEPY-UHFFFAOYSA-N
Synonym brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
PubChem CID 10458
ChEBI CHEBI:73718
IUPAC Name tridecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCCC(=O)O

Specifications

Linear Formula HO2C(CH2)11CO2H
Quantity 5 g
Formula Weight 244.33
Percent Purity 95%
Chemical Name or Material Undecane-1,11-dicarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.