missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic acid, 99%

Product Code. 10713181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713181 25 g Glass bottle 25g
10693363 5 g Glass bottle 5g
10655232 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10713181 Supplier Thermo Scientific Acros Supplier No. 173970250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-37-8 | C11H22O2 | 186.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-37-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.29
MDL Number MFCD00002730
InChI Key ZDPHROOEEOARMN-UHFFFAOYSA-N
Synonym hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
PubChem CID 8180
ChEBI CHEBI:32368
IUPAC Name undecanoic acid
SMILES CCCCCCCCCCC(=O)O

Specifications

Melting Point 28.5°C
Color White to Yellow
Boiling Point 284.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9CO2H
Quantity 25 g
Beilstein 02, 358
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,cresols and 1-butanol
Formula Weight 186.29
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Undecanoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.