missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Undecylenic acid, 98+%

Product Code. 10265480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10265480 250 g Glass bottle 250g
10317570 500 g Glass bottle 500g
10618064 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265480 Supplier Thermo Scientific Chemicals Supplier No. 154492500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00004442
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Synonym 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
PubChem CID 5634
ChEBI CHEBI:35045
IUPAC Name undec-10-enoic acid
SMILES C=CCCCCCCCCC(=O)O

Specifications

Melting Point 24.5°C
Color Yellow
Density 0.9110g/mL
Boiling Point 275.0°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8CO2H
Refractive Index 1.4470 to 1.4480
Quantity 250 g
Merck Index 15, 10033
Specific Gravity 0.911
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,benzene,with fixed and volatile oils
Formula Weight 184.28
Percent Purity 99%
Physical Form Crystalline Low Melting Solid or Liquid
Chemical Name or Material Undecylenic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.