Learn More
Köttermann Underbench Chemical Storage Cabinet
This lockable Underbench Chemical Storage Cabinet provides safe storage of aggressive non-flammable chemicals and toxic substances.
Brand:  Köttermann 375-0018
Additional Details : Weight : 25.00000kg
 View more versions of this product
Product Code. 17362149
  • 567.10 € / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • For the storage of non-corrosive, non-flammable substances
  • Can be integrated in modular cabinet assemblies
  • Lockable against unauthorized access
  • Capable of connecting to the exhaust air system for permanent ventilation
  • Double-walled, noise-insulated doors
Specifications
Specifications
516 x 600 x 720 mm
1 Right-hinged Door, 1 Shelf
Hinged, Locking
White
Non-corrosive, Non-flammable Chemical Storage
Steel (Galvanized)