Learn More
Köttermann Underbench Chemical Storage Cabinet
This lockable Underbench Chemical Storage Cabinet provides safe storage of aggressive non-flammable chemicals and toxic substances.
Brand:  Köttermann 375-0001
Additional Details : Weight : 77.68540kg
 View more versions of this product
Product Code. 17372139
  • 1293.20 € / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • For the storage of non-corrosive, non-flammable substances
  • Can be integrated in modular cabinet assemblies
  • Lockable against unauthorized access
  • Capable of connecting to the exhaust air system for permanent ventilation
  • Double-walled, noise-insulated doors
Specifications
Specifications
516 x 600 x 1920 mm
1 Left-hinged Door, 4 Shelves
Hinged, Locking
White
Non-corrosive, Non-flammable Chemical Storage
Steel (Galvanized)