Learn More
Köttermann Underbench Safety Cabinet
Product Code. 17392129
Change view
Click to view available options
Door Style:
Drawer
Unit Size:
Each
Product Code. Door Style unitSize
17392129 Drawer Each
1 options
Product Code. 17392129
Brand: Köttermann 37100032
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Underbench Safety Cabinet is ideal for flammable hazardous substances, complying with fire resistance class standards (Type 90).
  • Low design to fit under laboratory bench
  • Fire resistant (Type 90)
  • Features two pull-out doors
Specifications
Dimensions (D x W x H) 570 x 1400 x 720 mm
Door Style Drawer
No. of Doors 2