Learn More
Haldenwanger™ Unglazed Porcelain Pestle
Use these pestles for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Pestles are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 56/00/GLASIERT
Code : 02
Additional Details : Weight : 0.04860kg
 View more versions of this product
Product Code. 10656405
  • 58.45 € / Each
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Unglazed
Pestle
Porcelain
115mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products