Learn More
Hughes Unheated Emergency Safety Shower
Product Code. 17702358
Change view
Click to view available options
Material:
Galvanized Steel
Stainless Steel
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
17702358 Stainless Steel Each
17712358 Galvanized Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17702358 Supplier Hughes Supplier No. HSHS0018GS
  • ONLINE EXCLUSIVE PRICE
    1081.50 €
    823.66 € / Pack of 1
    Save 257.84 €
    23% Off
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Floor mounted unheated emergency safety shower, suitable for use in all applications.
  • Floor mounted unheated emergency safety shower
  • ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
  • Delivers the ANSI recommended 76 liters per minute of potable water
  • Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Material Stainless Steel
Mounting Floor
Product Type Safety Shower