Learn More
Abeba™ UNI6 36721 Shoes
Product Code. 15690458
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15690458 41 1 pair
15630458 35 1 pair
15640458 36 1 pair
15650458 37 1 pair
15660458 38 1 pair
15670458 39 1 pair
15680458 40 1 pair
15600468 42 1 pair
15610468 43 1 pair
15620468 44 1 pair
15630468 45 1 pair
15640468 46 1 pair
15650468 47 1 pair
15660468 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15690458 missing translation for 'mfr' Abeba™ missing translation for 'supplierNo' 36721/41
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
  • CE EN ISO 20347:2012 O1 FO SRC
  • Velours with breathable textile inlays
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O1 FO SRC
Color Black
Material Velour
Size 41
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36721